Assay Details
Gene Symbol: TRMT112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434522.1 | XP_015290008.1 | 1 - 2 | 245 | 92 | |
XM_015434523.1 | XP_015290009.1 | 1 - 2 | 642 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
