Assay Details
Gene Symbol: CEACAM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590121.2 | XP_005590178.1 | 3 - 4 | 808 | 91 | |
XM_015441933.1 | XP_015297419.1 | 3 - 4 | 808 | 91 | ||
XM_015441934.1 | XP_015297420.1 | 3 - 4 | 808 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
