Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319443.1 | NP_001306372.1 | 9 - 10 | 1028 | 77 | |
XM_005568655.2 | XP_005568712.1 | 10 - 11 | 1475 | 77 | ||
XM_015457902.1 | XP_015313388.1 | 10 - 11 | 1447 | 77 | ||
GenBank mRNA | AJ320181.1 | - | 9 - 10 | 1028 | 77 |
