Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319443.1 | NP_001306372.1 | 1 - 2 | 120 | 87 | |
XM_005568655.2 | XP_005568712.1 | 2 - 3 | 567 | 87 | ||
XM_015457902.1 | XP_015313388.1 | 2 - 3 | 539 | 87 | ||
GenBank mRNA | AJ320181.1 | - | 1 - 2 | 120 | 87 |
