Assay Details
Gene Symbol: RASSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578588.2 | XP_005578645.1 | 1 | 1654 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
