Assay Details
Gene Symbol: CDKN2AIPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557786.2 | XP_005557843.1 | 2 - 3 | 404 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
