Assay Details
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543910.2 | XP_005543967.1 | 3 - 4 | 3293 | 92 | |
XM_005543912.2 | XP_005543969.1 | 3 - 4 | 1268 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
