Assay Details
Gene Symbol: GPANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553508.2 | XP_005553565.1 | 3 - 4 | 1153 | 77 | |
XM_005553509.2 | XP_005553566.1 | 3 - 4 | 956 | 77 | ||
XM_005553510.2 | XP_005553567.1 | 3 - 4 | 959 | 77 | ||
XM_015449625.1 | XP_015305111.1 | 2 - 3 | 1227 | 77 | ||
XM_015449626.1 | XP_015305112.1 | 2 - 3 | 1459 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
