Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283681.1 | NP_001270610.1 | 9 - 10 | 1152 | 91 | |
XM_015452080.1 | XP_015307566.1 | 9 - 10 | 1298 | 91 | ||
XM_015452084.1 | XP_015307570.1 | 8 - 9 | 1254 | 91 | ||
GenBank mRNA | AB169307.1 | - | 9 - 10 | 1152 | 91 |
