Assay Details
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283398.1 | NP_001270327.1 | 6 - 7 | 780 | 100 | |
XM_015432111.1 | XP_015287597.1 | 5 - 6 | 503 | 100 | ||
XM_015432112.1 | XP_015287598.1 | 4 - 5 | 432 | 100 | ||
XM_015432113.1 | XP_015287599.1 | 3 - 4 | 322 | 100 | ||
XM_015432114.1 | XP_015287600.1 | 7 - 8 | 618 | 100 | ||
XM_015432115.1 | XP_015287601.1 | 5 - 6 | 436 | 100 | ||
GenBank mRNA | AB070144.1 | - | 6 - 7 | 781 | 100 |
