Assay Details
Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283651.1 | NP_001270580.1 | 3 - 4 | 269 | 92 | |
GenBank mRNA | AB174188.1 | - | 3 - 4 | 269 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
