Assay Details
Gene Symbol: NDUFS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284025.1 | NP_001270954.1 | 154 | 119 | ||
GenBank mRNA | AB169414.1 | - | 154 | 119 |

More Information
Set Membership: |
![]() |
Panther Classification:
