Assay Details
Gene Symbol: GHV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290290.1 | NP_001277219.1 | 4 - 5 | 497 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
