Assay Details
Gene Symbol: CAPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576185.2 | XP_005576242.1 | 10 - 12 | 1554 | 59 | |
XM_005576187.2 | XP_005576244.1 | 10 - 12 | 1554 | 59 | ||
XM_005576188.2 | XP_005576245.1 | 11 - 13 | 1419 | 59 | ||
XM_015433829.1 | XP_015289315.1 | 10 - 12 | 3511 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
