Assay Details
Gene Symbol: GET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548666.2 | XP_005548723.2 | 3 - 4 | 451 | 80 | |
XM_015446847.1 | XP_015302333.1 | 3 - 4 | 337 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
