Assay Details
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015437421.1 | XP_015292907.1 | 2 - 3 | 664 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
