Assay Details
Gene Symbol: PRSS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442399.1 | XP_015297885.1 | 3 - 4 | 596 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
