Assay Details
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557936.2 | XP_005557993.1 | 2 | 224 | 93 | |
XM_015451956.1 | XP_015307442.1 | 2 | 224 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005557936.2, XM_015451956.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
