Assay Details
Gene Symbol: Chrna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007389.5 | NP_031415.2 | 4 - 5 | 393 | 67 | |
GenBank mRNA | AK029177.1 | - | 4 - 5 | 362 | 67 | |
AK132589.1 | - | 4 - 5 | 333 | 67 | ||
BC125439.1 | - | 4 - 5 | 393 | 40129954 | 67 | |
BC137678.1 | - | 4 - 5 | 393 | 8860697 | 67 | |
X03986.1 | - | 4 - 5 | 350 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
