Assay Details
Gene Symbol: App | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198823.1 | NP_001185752.1 | 11 - 12 | 1605 | 130 | |
NM_001198824.1 | NP_001185753.1 | 10 - 11 | 1548 | 130 | ||
NM_001198825.1 | NP_001185754.1 | 10 - 11 | 1548 | 130 | ||
NM_001198826.1 | NP_001185755.1 | 11 - 12 | 1605 | 130 | ||
NM_007471.3 | NP_031497.2 | 9 - 10 | 1380 | 130 | ||
GenBank mRNA | AK029172.1 | - | 1 - 2 | 2773 | 130 | |
AK076506.1 | - | 3 - 4 | 300 | 130 | ||
AK128971.1 | - | 10 - 11 | 1557 | 130 | ||
AK133639.1 | - | 10 - 11 | 1545 | 130 | ||
AK147543.1 | - | 9 - 10 | 1416 | 130 | ||
AK148439.1 | - | 11 - 12 | 1478 | 130 | ||
AK159245.1 | - | 11 - 12 | 1614 | 130 | ||
AK159352.1 | - | 11 - 12 | 1611 | 130 | ||
AK159718.1 | - | 11 - 12 | 1591 | 130 | ||
AK161854.1 | - | 4 - 5 | 462 | 130 | ||
AK161976.1 | - | 9 - 10 | 1355 | 130 | ||
AY267348.1 | - | 11 - 12 | 1456 | 130 | ||
BC005490.1 | - | 7 - 8 | 969 | 130 | ||
BC070409.1 | - | 9 - 10 | 1376 | 5708093 | 130 | |
M18373.1 | - | 9 - 10 | 1369 | 130 | ||
U84012.1 | - | 9 - 10 | 1369 | 130 | ||
X59379.1 | - | 9 - 10 | 1299 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
