Assay Details
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037859.2 | NP_001032948.2 | 4 - 5 | 872 | 72 | |
GenBank mRNA | AK004947.1 | - | 3 - 4 | 682 | 72 | |
AK041299.1 | - | 3 - 4 | 660 | 72 | ||
AK076215.1 | - | 3 - 4 | 680 | 72 | ||
AK079247.1 | - | 3 - 4 | 689 | 72 | ||
AK080326.1 | - | 3 - 4 | 669 | 72 | ||
AK089498.1 | - | 3 - 4 | 662 | 72 | ||
AK143545.1 | - | 3 - 4 | 673 | 72 | ||
AK146069.1 | - | 4 - 5 | 873 | 72 | ||
AK154128.1 | - | 3 - 4 | 687 | 72 | ||
AK154556.1 | - | 3 - 4 | 683 | 72 | ||
AK154653.1 | - | 3 - 4 | 683 | 72 | ||
AK154699.1 | - | 3 - 4 | 683 | 72 | ||
AK155488.1 | - | 3 - 4 | 692 | 72 | ||
AK155565.1 | - | 3 - 4 | 702 | 72 | ||
AK155576.1 | - | 689 | 72 | |||
AK155640.1 | - | 3 - 4 | 687 | 72 | ||
AK155654.1 | - | 3 - 4 | 687 | 72 | ||
AK155858.1 | - | 3 - 4 | 690 | 72 | ||
AK156550.1 | - | 3 - 4 | 707 | 72 | ||
AK171241.1 | - | 3 - 4 | 722 | 72 | ||
AK171386.1 | - | 3 - 4 | 687 | 72 | ||
AK171543.1 | - | 3 - 4 | 662 | 72 | ||
BC036343.1 | - | 3 - 4 | 659 | 4948135 | 72 | |
BC043054.1 | - | 3 - 4 | 663 | 6418718 | 72 | |
BC050024.1 | - | 3 - 4 | 729 | 72 | ||
BC066863.1 | - | 4 - 5 | 781 | 30436119 | 72 | |
X06368.1 | - | 3 - 4 | 665 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
