Assay Details
Gene Symbol: Dnmt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271753.1 | NP_001258682.1 | 20 - 21 | 2658 | 80 | |
NM_007872.4 | NP_031898.1 | 20 - 21 | 2704 | 80 | ||
NM_153743.4 | NP_714965.1 | 15 - 16 | 1882 | 80 | ||
GenBank mRNA | AF068625.2 | - | 20 - 21 | 2608 | 80 | |
AF480164.1 | - | 15 - 16 | 1882 | 80 | ||
AK077792.1 | - | 15 - 16 | 1847 | 80 | ||
AK090132.1 | - | 20 - 21 | 2635 | 80 | ||
AK147263.1 | - | 20 - 21 | 2658 | 80 | ||
AK147627.1 | - | 20 - 21 | 2658 | 80 | ||
AK147642.1 | - | 20 - 21 | 2658 | 80 | ||
AK147676.1 | - | 20 - 21 | 2633 | 80 | ||
AK157792.1 | - | 15 - 16 | 1853 | 80 | ||
BC007466.1 | - | 20 - 21 | 2636 | 3492853 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
