Assay Details
Gene Symbol: Eef2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267710.1 | NP_001254639.1 | 12 - 13 | 1637 | 60 | |
NM_001267711.1 | NP_001254640.1 | 12 - 13 | 1632 | 60 | ||
NM_007908.4 | NP_031934.1 | 12 - 13 | 1721 | 60 | ||
GenBank mRNA | AK028524.1 | - | 12 - 13 | 1698 | 60 | |
AK080915.1 | - | 12 - 13 | 1638 | 60 | ||
AK136015.1 | - | 11 - 12 | 1363 | 60 | ||
BC003433.1 | - | 12 - 13 | 1669 | 2646503 | 60 | |
BC055361.1 | - | 12 - 13 | 1632 | 30046537 | 60 | |
U93848.1 | - | 11 - 12 | 1375 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
