Assay Details
Gene Symbol: Egfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007912.4 | NP_031938.1 | 9 - 10 | 1410 | 83 | |
NM_207655.2 | NP_997538.1 | 9 - 10 | 1410 | 83 | ||
GenBank mRNA | AF124513.1 | - | 9 - 10 | 1344 | 83 | |
AF275367.1 | - | 9 - 10 | 1353 | 83 | ||
AK004883.1 | - | 9 - 10 | 1411 | 83 | ||
AK004911.1 | - | 9 - 10 | 1385 | 83 | ||
AK004944.1 | - | 9 - 10 | 1404 | 83 | ||
AK134262.1 | - | 9 - 10 | 1383 | 83 | ||
BC023729.1 | - | 9 - 10 | 1381 | 5351807 | 83 | |
U03425.1 | - | 9 - 10 | 1180 | 83 | ||
X59698.1 | - | 9 - 10 | 1289 | 83 | ||
X78987.1 | - | 9 - 10 | 1189 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
