Assay Details
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB560752.1 | - | 2 - 3 | 669 | 99 | |
AB560753.1 | - | 1 - 2 | 532 | 99 | ||
AB560754.1 | - | 1 - 2 | 532 | 99 | ||
AB560755.1 | - | 1 - 2 | 532 | 99 | ||
AB560756.1 | - | 1 - 2 | 532 | 99 | ||
AK036627.1 | - | 2 - 3 | 643 | 99 | ||
AK039911.1 | - | 5 - 6 | 1146 | 99 | ||
AK041525.1 | - | 3 - 4 | 791 | 99 | ||
AK077236.1 | - | 2 - 3 | 648 | 99 | ||
AK087638.1 | - | 2 - 3 | 621 | 99 | ||
AK139745.1 | - | 3 - 4 | 1203 | 99 | ||
EU791538.1 | - | 2 - 3 | 587 | 99 | ||
EU791540.1 | - | 2 - 3 | 712 | 99 | ||
M38651.1 | - | 2 - 3 | 651 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
