Assay Details
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161373.1 | NP_001154845.1 | 14 - 15 | 5475 | 68 | |
NM_001161374.1 | NP_001154846.1 | 14 - 15 | 5475 | 68 | ||
NM_007977.2 | NP_032003.2 | 15 - 16 | 5685 | 68 | ||
GenBank mRNA | BC138347.1 | - | 14 - 15 | 5356 | 8861367 | 68 |
BC145245.1 | - | 14 - 15 | 5356 | 9053797 | 68 | |
L05573.1 | - | 15 - 16 | 5683 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
