Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 16 - 17 | 1705 | 86 | |
NM_007982.2 | NP_032008.2 | 15 - 16 | 1550 | 86 | ||
GenBank mRNA | AK079821.1 | - | 16 - 17 | 1802 | 86 | |
AK220543.1 | - | 15 - 16 | 1550 | 86 | ||
BC030180.1 | - | 16 - 17 | 1635 | 86 | ||
M95408.1 | - | 14 - 15 | 1347 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
