Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 20 - 21 | 2105 | 77 | |
NM_007982.2 | NP_032008.2 | 19 - 20 | 1950 | 77 | ||
GenBank mRNA | AB011499.1 | - | 20 - 21 | 1761 | 77 | |
AB030035.1 | - | 18 - 19 | 1576 | 77 | ||
AK079821.1 | - | 20 - 21 | 2202 | 77 | ||
AK220543.1 | - | 19 - 20 | 1950 | 77 | ||
BC030180.1 | - | 20 - 21 | 2035 | 77 | ||
JN971016.1 | - | 19 - 20 | 1677 | 77 | ||
M95408.1 | - | 18 - 19 | 1747 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
