Assay Details
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008011.2 | NP_032037.2 | 16 - 17 | 2309 | 132 | |
GenBank mRNA | AK084850.1 | - | 16 - 17 | 2309 | 132 | |
AK085723.1 | - | 16 - 17 | 2307 | 132 | ||
BC033313.1 | - | 16 - 17 | 2496 | 5132831 | 132 | |
DQ388428.1 | - | 15 - 16 | 2195 | 132 | ||
X59927.1 | - | 16 - 17 | 2278 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
