Assay Details
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF256197.1 | - | 8 - 9 | 98 | ||
AF256198.1 | - | 8 - 9 | 98 | |||
X51986.1 | - | 8 - 9 | 1293 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
