Assay Details
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008072.2 | NP_032098.2 | 1 - 2 | 170 | 103 | |
GenBank mRNA | BC116832.1 | - | 1 - 2 | 105 | 40126151 | 103 |
BC116836.1 | - | 1 - 2 | 105 | 40126155 | 103 | |
S42882.1 | - | 1 - 2 | 163 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC116832.1, BC116836.1, NM_008072.2, S42882.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Altered GABAA receptor-mediated synaptic transmission disrupts the firing of gonadotropin-releasing hormone neurons in male mice under conditions that mimic steroid abuse.
- Authors
- Penatti CA, Davis MC, Porter DM, Henderson LP
- Journal
- J Neurosci - Page No 6497 (Wed May 12 00:00:00 UTC 2010)
- Volume (Year)
- 30 (2010)
- Product Usage
- -
