Assay Details
Gene Symbol: Gpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008156.2 | NP_032182.2 | 3 - 4 | 460 | 74 | |
GenBank mRNA | AF050666.1 | - | 3 - 4 | 414 | 74 | |
AK004957.2 | - | 2 - 3 | 164 | 74 | ||
AY081194.1 | - | 2 - 3 | 187 | 74 | ||
AY333960.1 | - | 2 - 3 | 144 | 74 | ||
BC019146.1 | - | 2 - 3 | 173 | 5052822 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
