Assay Details
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008170.2 | NP_032196.2 | 7 - 8 | 1776 | 121 | |
GenBank mRNA | D10217.2 | - | 7 - 8 | 1776 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
