Assay Details
Gene Symbol: Il12rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008353.2 | NP_032379.2 | 12 - 13 | 1653 | 82 | |
GenBank mRNA | AK137577.1 | - | 12 - 13 | 1654 | 82 | |
AK171937.1 | - | 12 - 13 | 1627 | 82 | ||
BC057212.1 | - | 12 - 13 | 1636 | 5350447 | 82 | |
U23922.1 | - | 12 - 13 | 1608 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
