Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008369.1 | NP_032395.1 | 6 - 7 | 836 | 74 | |
GenBank mRNA | BC049889.1 | - | 5 - 6 | 1236 | 74 | |
FJ550346.1 | - | 3 - 4 | 418 | 74 | ||
X64534.1 | - | 6 - 7 | 836 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
