Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008369.1 | NP_032395.1 | 9 - 10 | 1107 | 105 | |
GenBank mRNA | BC049889.1 | - | 8 - 9 | 1507 | 105 | |
FJ550346.1 | - | 6 - 7 | 689 | 105 | ||
X64534.1 | - | 9 - 10 | 1107 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
