Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008369.1 | NP_032395.1 | 11 - 12 | 1273 | 79 | |
GenBank mRNA | BC049889.1 | - | 10 - 11 | 1673 | 79 | |
BC077736.1 | - | 5 - 6 | 614 | 79 | ||
FJ550346.1 | - | 8 - 9 | 855 | 79 | ||
X64534.1 | - | 11 - 12 | 1273 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008181 | X64534.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008181 | BC049889.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008181 | BC077736.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008181 | NM_008369.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313123 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203284 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814248 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156232 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768480 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150462 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122107 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008181 | X64534.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008181 | BC049889.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008181 | BC077736.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008181 | NM_008369.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432651 | X64534.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432651 | BC049889.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432651 | FJ550346.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432651 | NM_008369.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29420289 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08034782 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122107 | - | - |
Whole Mouse Genome G4122A | A_51_P432651 | X64534.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P432651 | BC049889.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P432651 | FJ550346.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P432651 | NM_008369.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115035 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122107 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11550459 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11550478 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11550456 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11550480 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11550453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29420299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11550463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11743831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11550462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11550459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11550493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11550456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11550467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29420276 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10412766 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412764 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412761 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412767 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412768 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412770 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412765 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412763 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412762 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412769 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419712_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10412760 | - | - |
Mouse Genome 430 2.0 Array | 1419712_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419712_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16188_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16188_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400000911.mm.1 | TC1400000112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400000911.mm.1 | NM_008369.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400000911.mm.1 | NM_008369.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400000910.mm.1 | TC1400000112.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400000910.mm.1 | NM_008369.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400000910.mm.1 | NM_008369.1 | Refseq |
Mouse Expression Set 430 | 1419712_PM_at | - | - |
Mouse Expression Set 430 | 1419712_at | - | - |
Murine Genome U74v2 Set | 92955_PM_at | - | - |
Murine Genome U74v2 Set | 92955_at | - | - |
Mouse Genome 430A 2.0 Array | 1419712_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419712_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419712_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419712_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 446448 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6420722 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420722 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724449 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2724449 | - | - |
mouse-6 v1.1 (Illumina) | scl46617.10.1_4-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420722 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46617.10.1_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420722 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2916571 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2916571 | - | - |
