Assay Details
Gene Symbol: Il9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008373.1 | NP_032399.1 | 3 - 4 | 194 | 85 | |
GenBank mRNA | X14045.1 | - | 3 - 4 | 194 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
