Assay Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008468.4 | NP_032494.3 | 3 - 4 | 317 | 72 | |
GenBank mRNA | AF020773.1 | - | 2 - 3 | 221 | 72 | |
AK046371.1 | - | 3 - 4 | 276 | 72 | ||
AK084212.1 | - | 3 - 4 | 275 | 72 | ||
BC004833.1 | - | 2 - 3 | 233 | 2651272 | 72 | |
CJ082540.1 | - | 3 - 4 | 338 | 72 | ||
CT010260.1 | - | 2 - 3 | 221 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
