Assay Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008468.4 | NP_032494.3 | 4 - 5 | 422 | 96 | |
GenBank mRNA | AF020773.1 | - | 3 - 4 | 326 | 96 | |
AK046371.1 | - | 4 - 5 | 381 | 96 | ||
AK084212.1 | - | 4 - 5 | 380 | 96 | ||
AK154882.1 | - | 4 - 5 | 407 | 96 | ||
BC004833.1 | - | 3 - 4 | 338 | 2651272 | 96 | |
CT010260.1 | - | 3 - 4 | 326 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
