Assay Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008468.4 | NP_032494.3 | 12 - 13 | 1331 | 89 | |
GenBank mRNA | AF020773.1 | - | 11 - 12 | 1235 | 89 | |
AK007053.1 | - | 1 - 2 | 101 | 89 | ||
AK046371.1 | - | 12 - 13 | 1290 | 89 | ||
AK084212.1 | - | 12 - 13 | 1289 | 89 | ||
AK154882.1 | - | 12 - 13 | 1316 | 89 | ||
BC004833.1 | - | 11 - 12 | 1247 | 2651272 | 89 | |
CT010260.1 | - | 11 - 12 | 1235 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
