Assay Details
Gene Symbol: Ltk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008523.2 | NP_032549.2 | 8 - 9 | 1107 | 103 | |
NM_203345.2 | NP_976220.2 | 14 - 15 | 1953 | 103 | ||
NM_206941.1 | NP_996824.1 | 9 - 10 | 1290 | 103 | ||
NM_206942.2 | NP_996825.2 | 13 - 14 | 1770 | 103 | ||
GenBank mRNA | AK157478.1 | - | 6 - 7 | 1104 | 103 | |
BC047138.1 | - | 8 - 9 | 1107 | 6389834 | 103 | |
BC141299.1 | - | 13 - 14 | 1746 | 9055911 | 103 | |
M90470.1 | - | 14 - 15 | 1953 | 103 | ||
X07984.1 | - | 8 - 9 | 1081 | 103 | ||
X52621.1 | - | 8 - 9 | 1096 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
