Assay Details
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 11 - 12 | 1725 | 53 | |
GenBank mRNA | L11625.1 | - | 2 - 3 | 263 | 53 | |
U21301.1 | - | 11 - 12 | 1725 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
