Assay Details
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 14 - 15 | 1997 | 66 | |
GenBank mRNA | AK029009.1 | - | 3 - 4 | 199 | 66 | |
AK030158.1 | - | 6 - 7 | 696 | 66 | ||
L11625.1 | - | 5 - 6 | 535 | 66 | ||
U21301.1 | - | 14 - 15 | 1997 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
