Assay Details
Gene Symbol: Gadd45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008655.1 | NP_032681.1 | 1 - 2 | 275 | 53 | |
GenBank mRNA | AK010420.1 | - | 1 - 2 | 293 | 53 | |
AK149895.1 | - | 1 - 2 | 293 | 53 | ||
AK154952.1 | - | 1 - 2 | 293 | 53 | ||
BC023815.1 | - | 1 - 2 | 287 | 5355150 | 53 | |
X54149.1 | - | 1 - 2 | 275 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
