Assay Details
Gene Symbol: Slc11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146161.1 | NP_001139633.1 | 4 - 5 | 444 | 64 | |
NM_008732.2 | NP_032758.2 | 4 - 5 | 444 | 64 | ||
GenBank mRNA | AF029758.1 | - | 4 - 5 | 379 | 64 | |
AK049856.1 | - | 4 - 5 | 392 | 64 | ||
AK083478.1 | - | 4 - 5 | 390 | 64 | ||
AK148276.1 | - | 4 - 5 | 392 | 64 | ||
AK210047.1 | - | 3 - 4 | 315 | 64 | ||
BC019137.1 | - | 4 - 5 | 418 | 5040976 | 64 | |
L33415.1 | - | 4 - 5 | 372 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
