Assay Details
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162415.1 | NP_001155887.1 | 6 - 7 | 606 | 85 | |
NM_001162416.1 | NP_001155888.1 | 4 - 5 | 399 | 85 | ||
NM_008825.4 | NP_032851.2 | 5 - 6 | 500 | 85 | ||
NR_027859.1 | 5 - 6 | 500 | 85 | |||
GenBank mRNA | AK029659.1 | - | 6 - 7 | 607 | 85 | |
AK146744.1 | - | 5 - 6 | 494 | 85 | ||
BC018418.1 | - | 5 - 6 | 457 | 3979400 | 85 | |
BC051014.1 | - | 4 - 5 | 413 | 6493944 | 85 | |
EU616666.1 | - | 4 - 5 | 398 | 85 | ||
EU616667.1 | - | 4 - 5 | 398 | 85 | ||
EU616668.1 | - | 4 - 5 | 398 | 85 | ||
EU616669.1 | - | 4 - 5 | 398 | 85 | ||
EU616670.1 | - | 4 - 5 | 408 | 85 | ||
X98847.1 | - | 4 - 5 | 418 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
