Assay Details
Gene Symbol: Pfkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008826.4 | NP_032852.2 | 9 - 10 | 1047 | 59 | |
GenBank mRNA | AK030511.1 | - | 9 - 10 | 1002 | 59 | |
AK030892.1 | - | 2 - 3 | 146 | 59 | ||
AK036318.1 | - | 9 - 10 | 980 | 59 | ||
AK081313.1 | - | 9 - 10 | 998 | 59 | ||
AK159328.1 | - | 9 - 10 | 992 | 59 | ||
AK168660.1 | - | 9 - 10 | 1003 | 59 | ||
BC020097.1 | - | 9 - 10 | 1001 | 4014395 | 59 | |
BC029336.1 | - | 9 - 10 | 991 | 59 | ||
J03928.1 | - | 9 - 10 | 976 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK030892.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
