Assay Details
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008839.2 | NP_032865.2 | 17 - 18 | 2718 | 62 | |
GenBank mRNA | AK051885.1 | - | 17 - 18 | 2351 | 62 | |
BC089038.1 | - | 17 - 18 | 2718 | 30692343 | 62 | |
BC130228.1 | - | 17 - 18 | 2667 | 40141870 | 62 | |
U03279.1 | - | 17 - 18 | 2663 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
