Assay Details
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008941.3 | NP_032967.1 | 16 - 17 | 1872 | 99 | |
NM_178855.4 | NP_849186.2 | 15 - 16 | 1827 | 99 | ||
GenBank mRNA | AK033582.1 | - | 15 - 16 | 1829 | 99 | |
AK038356.1 | - | 7 - 8 | 821 | 99 | ||
BC117917.1 | - | 16 - 17 | 1871 | 40097367 | 99 | |
BC117918.1 | - | 16 - 17 | 1871 | 40097370 | 99 | |
U73378.1 | - | 16 - 17 | 1867 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
